2,4-Dibromopyrimidine 95%

Supplier: Ambeed
Warning

A126335-10G A126335-1G A126335-250MG A126335-25G A126335-5G
76766-888EA 198 USD
76766-888 76766-890 76766-892 76766-894 76766-896
2,4-Dibromopyrimidine 95%
2,4-Dibromopyrimidine

Precaución: for Research Use Only

Formula: C₄H₂Br₂N₂
MDL Number: MFCD06798232
CAS Number: 3921-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR