6-Bromo-2,3-difluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4-difluoro-2-methylbenzene

A125868-1G A125868-250MG
76766-876EA 25.47 USD
76766-876 76766-878
6-Bromo-2,3-difluorotoluene 98%
6-Bromo-2,3-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD07777161
CAS Number: 847502-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR