6-Chloro-L-Tryptophan 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid, H-Trp(6-Cl)-OH

A124813-100MG A124813-1G A124813-250MG
76766-836EA 27 USD
76766-836 76766-838 76766-840
6-Chloro-L-Tryptophan 98%
6-Chloro-L-Tryptophan

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.7 g/mol
MDL Number: MFCD02683925
CAS Number: 33468-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR