4-Methyl-N,N-diphenylaniline 98%

Supplier: Ambeed
Warning

A124538-100G A124538-25G A124538-5G
76766-810EA 119 USD
76766-810 76766-812 76766-814
4-Methyl-N,N-diphenylaniline 98%
4-Methyl-N,N-diphenylaniline

Precaución: for Research Use Only

Formula: C₁₉H₁₇N
MDL Number: MFCD03093256
CAS Number: 4316-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR