5-Amino-2-nitropyridine 98%

Supplier: Ambeed
Warning

A124465-1G A124465-250MG A124465-5G A124465-10G A124465-100MG
76766-802EA 20 USD
76766-802 76766-804 77571-468 77624-042 77876-899
5-Amino-2-nitropyridine 98%
5-Amino-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MDL Number: MFCD01646117
CAS Number: 14916-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR