2-Fluoro-6-nitrotoluene 98%

Supplier: Ambeed
Warning

A124321-100G A124321-25G A124321-5G
76766-794EA 33 USD
76766-794 76766-796 76766-798 76766-800
2-Fluoro-6-nitrotoluene 98%
2-Fluoro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD00007197
CAS Number: 769-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR