5-Oxo-1-phenylpyrrolidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A124297-100MG A124297-1G A124297-250MG A124297-5G
76766-786EA 15 USD
76766-786 76766-788 76766-790 76766-792
5-Oxo-1-phenylpyrrolidine-3-carboxylic acid 98%
5-Oxo-1-phenylpyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00207656
CAS Number: 39629-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR