4-Bromo-3-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-methyl-2-nitrobenzene

A124225-10G A124225-25G A124225-5G
76766-780EA 11 USD
76766-780 76766-782 76766-784
4-Bromo-3-nitrotoluene 98%
4-Bromo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 35
MDL Number: MFCD00024180
CAS Number: 5326-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR