5-Bromo-2,4-dichloropyridine 98%

Supplier: Ambeed
Warning

A124118-100G A124118-10G A124118-1G A124118-25G A124118-5G
76766-768EA 498.63 USD
76766-768 76766-770 76766-772 76766-774 76766-776
5-Bromo-2,4-dichloropyridine 98%
5-Bromo-2,4-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MDL Number: MFCD09031035
CAS Number: 849937-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR