5-(4-Fluorophenyl)oxazole 97%

Supplier: Ambeed
Warning

A123924-10G A123924-1G
76766-764EA 425 USD
76766-764 76766-766
5-(4-Fluorophenyl)oxazole 97%
5-(4-Fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆FNO
MDL Number: MFCD00085087
CAS Number: 128101-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR