(2,4-Dimethyl-1,3-thiazol-5-yl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethyl-5-(hydroxymethyl)-1,3-thiazole

A123688-1G A123688-250MG A123688-5G
76766-758EA 67.9 USD
76766-758 76766-760 76766-762
(2,4-Dimethyl-1,3-thiazol-5-yl)methanol 98%
(2,4-Dimethyl-1,3-thiazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₉NOS
MW: 143.21 g/mol
MDL Number: MFCD03086098
CAS Number: 50382-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR