5-Bromo-3-methylbenzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A123533-10G A123533-1G A123533-250MG A123533-25G A123533-5G
76766-734EA 87 USD
76766-734 76766-736 76766-738 76766-740 76766-742
5-Bromo-3-methylbenzene-1,2-diamine 98%
5-Bromo-3-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.0600 g/mol
MDL Number: MFCD06411036
CAS Number: 76153-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR