6-Fluoroisatin 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1H-indole-2,3-dione, 6-Fluoroindoline-2,3-dione

A123527-100G A123527-1G A123527-5G A123527-25G A123527-10G
76766-728EA 352.22 USD
76766-728 76766-730 76766-732 77571-432 77592-410
6-Fluoroisatin 97%
6-Fluoroisatin

Precaución: for Research Use Only

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
MDL Number: MFCD03618556
CAS Number: 324-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR