2,5-Dibromo-3-hexylthiophene 98%

Supplier: Ambeed
Warning

A123133-1G A123133-25G A123133-5G
76766-692EA 37.14 USD
76766-692 76766-694 76766-696 76766-698
2,5-Dibromo-3-hexylthiophene 98%
2,5-Dibromo-3-hexylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₄Br₂S
MDL Number: MFCD00274313
CAS Number: 116971-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR