Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chloro-2-trifluoromethylpyrimidine-5-carboxylate

A123091-1G A123091-250MG
76766-686EA 23.34 USD
76766-686 76766-688
Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate 98%
Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆ClF₃N₂O₂
MW: 254.5937 g/mol
MDL Number: MFCD00173897
CAS Number: 720-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR