2,5-Dibromo-3-butylthiophene 97%

Supplier: Ambeed
Warning

A122837-1G A122837-250MG A122837-25G A122837-5G
76766-650EA 8.49 USD
76766-650 76766-652 76766-654 76766-656
2,5-Dibromo-3-butylthiophene 97%
2,5-Dibromo-3-butylthiophene

Precaución: for Research Use Only

Formula: C₈H₁₀Br₂S
MW: 298.04 g/mol
MDL Number: MFCD00672012
CAS Number: 116971-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR