(5-Bromo-1,3-phenylene)dimethanol 95%

Supplier: Ambeed
Warning

A122781-100MG A122781-1G A122781-250MG
76766-636EA 45.62 USD
76766-636 76766-638 76766-640
(5-Bromo-1,3-phenylene)dimethanol 95%
(5-Bromo-1,3-phenylene)dimethanol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.05 g/mol
MDL Number: MFCD00229250
CAS Number: 51760-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR