5-Chloro-2-pentanone 97% stabilised

Supplier: Ambeed
Warning

Synonyms: 5-Chloropentan-2-one

Stabilization: Stabilised with potassium carbonate

A122716-100G A122716-25G A122716-500G A122716-5G
76766-614EA 26 USD
76766-614 76766-616 76766-618 76766-620
5-Chloro-2-pentanone 97% stabilised
5-Chloro-2-pentanone

Precaución: for Research Use Only

Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 71
Melting Pt: -48
MDL Number: MFCD00001008
CAS Number: 5891-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR