9,10-Phenanthrenequinone 98%

Supplier: Ambeed
Warning

A122675-100G A122675-10G A122675-1G A122675-25G A122675-500G A122675-5G
76766-600EA 69 USD
76766-600 76766-602 76766-604 76766-606 76766-608 76766-610
9,10-Phenanthrenequinone 98%
9,10-Phenanthrenequinone

Precaución: for Research Use Only

Formula: C₁₄H₈O₂
MDL Number: MFCD00001163
CAS Number: 84-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR