6-Methyl-4-trifluoromethyl-2(1H)-pyridone 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

A122580-100MG A122580-250MG A122580-5G A122580-1G
76766-590EA 20.16 USD
76766-590 76766-592 76766-594 77624-016
6-Methyl-4-trifluoromethyl-2(1H)-pyridone 95%
6-Methyl-4-trifluoromethyl-2(1H)-pyridone

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MW: 177.126 g/mol
MDL Number: MFCD00042479
CAS Number: 22123-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR