Diethyl cyclobutane-1,1-dicarboxylate 98%

Supplier: Ambeed
Warning

A122511-100G A122511-10G A122511-25G A122511-500G A122511-5G
76766-580EA 44.56 USD
76766-580 76766-582 76766-584 76766-586 76766-588
Diethyl cyclobutane-1,1-dicarboxylate 98%
Diethyl cyclobutane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.24 g/mol
MDL Number: MFCD00019261
CAS Number: 3779-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR