(6-Aminopyridin-3-yl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-(carboxymethyl)pyridine, 2-(6-Aminopyridin-3-yl)ethanoic acid
A122143-100MG
A122143-1G
A122143-250MG
76766-572EA
77
USD
76766-572
76766-574
76766-576
(6-Aminopyridin-3-yl)acetic acid 95%
(6-Aminopyridin-3-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ |
MDL Number:
MFCD09757480 CAS Number: 39658-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...