PDE5-IN-7 95%

Supplier: Ambeed
Warning

Synonyms: 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one, 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone

A122141-100G A122141-5G A122141-25G
76766-568EA 118.83 USD
76766-568 76766-570 77624-014
PDE5-IN-7 95%
PDE5-IN-7

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₄O₂
MW: 312.37 g/mol
MDL Number: MFCD03093929
CAS Number: 139756-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR