1-Bromo-3-(methylsulfonyl)benzene 98%

Supplier: Ambeed
Warning

A122139-1G A122139-25G A122139-5G
76766-562EA 12.74 USD
76766-562 76766-564 76766-566
1-Bromo-3-(methylsulfonyl)benzene 98%
1-Bromo-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
MDL Number: MFCD21364720
CAS Number: 34896-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR