5-Methyl-6-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A122071-10G A122071-1G A122071-25G A122071-5G
76766-550EA 54 USD
76766-550 76766-552 76766-554 76766-556
5-Methyl-6-nitro-1H-indazole 97%
5-Methyl-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD09026996
CAS Number: 72521-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR