5-Chloro-4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A121998-10G A121998-1G A121998-250MG A121998-5G
76766-544EA 21.22 USD
76766-544 76766-546 76766-548 77596-414
5-Chloro-4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%
5-Chloro-4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₂
MW: 184.58 g/mol
MDL Number: MFCD07368155
CAS Number: 147619-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR