Z-D-Dap-OH 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-Dap-OH

A121002-100G A121002-10G A121002-1G A121002-250MG A121002-25G A121002-5G
76766-472EA 515.6 USD
76766-472 76766-474 76766-476 76766-478 76766-480 77624-002
Z-D-Dap-OH 97%
Z-D-Dap-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MW: 238.243 g/mol
MDL Number: MFCD00237346
CAS Number: 62234-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR