3-(2-Furyl)acrylic acid 95%

Supplier: Ambeed
Warning

A120682-100G A120682-10G A120682-25G A120682-5G A120682-500G
76766-440EA 96 USD
76766-440 76766-442 76766-444 76766-446 77596-244
3-(2-Furyl)acrylic acid 95%
3-(2-Furyl)acrylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₃
MDL Number: MFCD00003257
CAS Number: 539-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR