6-Chloro-2-methoxypyrimidin-4-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-chloro-2-methoxypyrimidine

A120645-100MG A120645-1G A120645-250MG A120645-5G
76766-432EA 24.41 USD
76766-432 76766-434 76766-436 76766-438
6-Chloro-2-methoxypyrimidin-4-amine 95%
6-Chloro-2-methoxypyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O
MW: 159.5700 g/mol
MDL Number: MFCD11053596
CAS Number: 3286-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR