Benzenesulfonamide 98%

Supplier: Ambeed
Warning

A119509-100G A119509-25G A119509-500G A119509-1000G
76766-392EA 8.49 USD
76766-392 76766-394 76766-396 77623-994
Benzenesulfonamide 98%
Benzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
MDL Number: MFCD00007930
CAS Number: 98-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR