3-Fluoro-4-iodobenzotrifluoride 97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A119276-100MG A119276-1G A119276-250MG A119276-5G A119276-25G
76766-370EA 18 USD
76766-370 76766-372 76766-374 76766-376 77876-870
3-Fluoro-4-iodobenzotrifluoride 97% stabilized
3-Fluoro-4-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄I
MDL Number: MFCD07771984
CAS Number: 132554-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR