2-Bromo-α,α,α-trifluoro-5-nitrotoluene 98%

Supplier: Ambeed
Warning

A119032-25G A119032-10G
76766-338EA 14 USD
76766-338 77876-866
2-Bromo-α,α,α-trifluoro-5-nitrotoluene 98%
2-Bromo-α,α,α-trifluoro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD00014707
CAS Number: 367-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR