6-Bromo-3-nitroimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

A118526-1G A118526-250MG
76766-312EA 31 USD
76766-312 76766-314
6-Bromo-3-nitroimidazo[1,2-a]pyridine 98%
6-Bromo-3-nitroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD09864963
CAS Number: 64064-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR