6-Methoxy-2(3H)-benzoxazolone 98%

Supplier: Ambeed
Warning

Synonyms: Coixol, 6-Methoxybenzo[d]oxazol-2(3H)-one

A118406-1G A118406-250MG
76766-302EA 206.88 USD
76766-302 76766-304
6-Methoxy-2(3H)-benzoxazolone 98%
6-Methoxy-2(3H)-benzoxazolone

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.15 g/mol
MDL Number: MFCD00051726
CAS Number: 532-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR