1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A118358-100G
A118358-1G
A118358-250MG
A118358-25G
A118358-5G
76766-292EA
199.45
USD
76766-292
76766-294
76766-296
76766-298
76766-300
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 98%
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.1400 g/mol |
MDL Number:
MFCD00159636 CAS Number: 5744-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...