3-Hydroxy-4-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A117757-100G A117757-10G A117757-25G A117757-500G A117757-5G
76766-282EA 22 USD
76766-282 76766-284 76766-286 76766-288 76766-290
3-Hydroxy-4-nitrobenzoic acid 98%
3-Hydroxy-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MDL Number: MFCD00007110
CAS Number: 619-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR