6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A117372-100MG A117372-1G A117372-250MG A117372-5G
76766-270EA 15.92 USD
76766-270 76766-272 76766-274 76766-276
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid 97%
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MDL Number: MFCD04974055
CAS Number: 14714-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR