5,7-Dihydroxycoumarin 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroxy-2H-chromen-2-one

A117068-100MG A117068-1G A117068-250MG A117068-5G
76766-262EA 11.67 USD
76766-262 76766-264 76766-266 76766-268
5,7-Dihydroxycoumarin 98%
5,7-Dihydroxycoumarin

Precaución: for Research Use Only

Formula: C₉H₆O₄
MW: 178.142 g/mol
MDL Number: MFCD00488354
CAS Number: 2732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR