6-Nitro-2-benzothiazolinone 97%

Supplier: Ambeed
Warning

A117049-1G A117049-250MG A117049-5G
76766-256EA 26.53 USD
76766-256 76766-258 76766-260
6-Nitro-2-benzothiazolinone 97%
6-Nitro-2-benzothiazolinone

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃S
MW: 196.183 g/mol
MDL Number: MFCD00239350
CAS Number: 28620-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR