2-Amino-5,6-dichlorobenzimidazole 96%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-1H-benzo[d]imidazol-2-amine

A116882-100MG A116882-1G A116882-250MG A116882-5G
76766-250EA 33.95 USD
76766-250 76766-252 76766-254 77596-020
2-Amino-5,6-dichlorobenzimidazole 96%
2-Amino-5,6-dichlorobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
MW: 202.04 g/mol
MDL Number: MFCD08234602
CAS Number: 18672-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR