4-Chloro-2-fluorobenzonitrile 98%

Supplier: Ambeed
Danger

A116866-10G A116866-1G A116866-25G
76766-236EA 15 USD
76766-236 76766-238 76766-240
4-Chloro-2-fluorobenzonitrile 98%
4-Chloro-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClFN
MDL Number: MFCD00143284
CAS Number: 57381-51-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR