Cytidine-5'-monophosphate disodium salt hydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5'-Cytidylic Acid Disodium Salt, 5'-CMP 2Na
A116088-100G
A116088-10G
A116088-25G
A116088-500G
A116088-5G
76766-184EA
30
USD
76766-184
76766-186
76766-188
76766-190
76766-192
Cytidine-5'-monophosphate disodium salt hydrate 97%
Cytidine-5'-monophosphate disodium salt hydrate
Precaución: for Research Use Only
Formula:
C₉H₁₄N₃O₈P MW: 367.16 g/mol |
MDL Number:
MFCD00064355 CAS Number: 6757-06-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...