2-Fluoro-5-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Fluor-2-hydroxy-4-(trifluormethyl)benzol, 2-Fluoro-5-trifluoromethylphenol

A116002-10G A116002-1G A116002-25G A116002-5G
76766-176EA 18.04 USD
76766-176 76766-178 76766-180 76766-182
2-Fluoro-5-(trifluoromethyl)phenol 98%
2-Fluoro-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₄O
MW: 180.1 g/mol
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR