5-Phenyl-2-thiopheneboronic acid 95%

Supplier: Ambeed
Warning

A115909-1G A115909-250MG A115909-5G A115909-25G
76766-170EA 35 USD
76766-170 76766-172 76766-174 77876-856
5-Phenyl-2-thiopheneboronic acid 95%
5-Phenyl-2-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BO₂S
MDL Number: MFCD01114643
CAS Number: 306934-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR