6-Chloro-2-phenylimidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A115784-100MG A115784-10G A115784-1G A115784-250MG A115784-5G
76766-162EA 6 USD
76766-162 76766-164 76766-166 76766-168 77876-853
6-Chloro-2-phenylimidazo[1,2-a]pyridine 95%
6-Chloro-2-phenylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂
MDL Number: MFCD00269393
CAS Number: 168837-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR