4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A115771-1G A115771-250MG A115771-5G A115771-25G
76766-156EA 7 USD
76766-156 76766-158 76766-160 77623-966
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₄N
MW: 258.01 g/mol
MDL Number: MFCD04973757
CAS Number: 104460-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR