2-Amino-3-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

A115177-100G A115177-10G A115177-1G A115177-25G A115177-5G
76766-106EA 364 USD
76766-106 76766-108 76766-110 76766-112 76766-114
2-Amino-3-fluorobenzonitrile 97%
2-Amino-3-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MDL Number: MFCD07368774
CAS Number: 115661-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR