6-Nitroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A114991-10G A114991-1G A114991-250MG A114991-25G A114991-5G A114991-100G
76766-096EA 77.45 USD
76766-096 76766-098 76766-100 76766-102 76766-104 77623-964
6-Nitroimidazo[1,2-a]pyridine 97%
6-Nitroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
MDL Number: MFCD05863333
CAS Number: 25045-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR