Ethyl-3-(trifluoromethyl)benzoate 97%
Supplier: Ambeed
|
Warning
|
A114339-100G
A114339-10G
A114339-1G
A114339-25G
A114339-5G
76766-050EA
103
USD
76766-050
76766-052
76766-054
76766-056
76766-058
Ethyl-3-(trifluoromethyl)benzoate 97%
Ethyl-3-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O₂ |
MDL Number:
MFCD00013560 CAS Number: 76783-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...