(5-Methyl-1H-indol-3-yl)methanol 95%

Supplier: Ambeed
Warning

A113077-100MG A113077-1G A113077-250MG
76766-006EA 60.48 USD
76766-006 76766-008 76766-010
(5-Methyl-1H-indol-3-yl)methanol 95%
(5-Methyl-1H-indol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.20 g/mol
MDL Number: MFCD06202775
CAS Number: 215997-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR